Assay Details
| Gene Symbol: ek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131095.1 | NP_571170.1 | 15 - 16 | 2824 | 84 | |
| GenBank mRNA | AJ005028.1 | - | 9 - 10 | 1226 | 84 | |
| BC162500.1 | - | 15 - 16 | 2740 | 84 | ||
| U89295.1 | - | 15 - 16 | 2824 | 84 | ||
More Information
Set Membership: |
|

ek1
Genomic Map