Assay Details
| Gene Symbol: ek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131095.1 | NP_571170.1 | 6 - 7 | 1565 | 91 | |
| GenBank mRNA | KM655625.1 | - | 6 - 7 | 1440 | 91 | |
| KM655626.1 | - | 6 - 7 | 1440 | 91 | ||
| U89295.1 | - | 6 - 7 | 1565 | 91 | ||
More Information
Set Membership: |
|

ek1
Genomic Map