Assay Details
| Gene Symbol: ek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131095.1 | NP_571170.1 | 7 - 8 | 1731 | 125 | |
| GenBank mRNA | AJ005028.1 | - | 1 - 2 | 133 | 125 | |
| BC162500.1 | - | 7 - 8 | 1647 | 125 | ||
| U89295.1 | - | 7 - 8 | 1731 | 125 | ||
More Information
Set Membership: |
|

ek1
Genomic Map