Assay Details
| Gene Symbol: ek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131095.1 | NP_571170.1 | 12 - 13 | 2277 | 76 | |
| GenBank mRNA | AJ005028.1 | - | 6 - 7 | 679 | 76 | |
| U89295.1 | - | 12 - 13 | 2277 | 76 | ||
More Information
Set Membership: |
|

ek1
Genomic Map