Assay Details
| Gene Symbol: mef2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131301.2 | NP_571376.2 | 3 - 4 | 522 | 61 | |
| XM_005159042.4 | XP_005159099.1 | 3 - 4 | 531 | 61 | ||
| XM_005159044.4 | XP_005159101.1 | 3 - 4 | 531 | 61 | ||
| XM_005159045.4 | XP_005159102.1 | 3 - 4 | 531 | 61 | ||
| XM_021467574.1 | XP_021323249.1 | 3 - 4 | 532 | 61 | ||
| XM_021467575.1 | XP_021323250.1 | 3 - 4 | 533 | 61 | ||
| XM_021467576.1 | XP_021323251.1 | 3 - 4 | 532 | 61 | ||
| XM_021467577.1 | XP_021323252.1 | 3 - 4 | 532 | 61 | ||
| XM_021467578.1 | XP_021323253.1 | 3 - 4 | 532 | 61 | ||
| XM_021467579.1 | XP_021323254.1 | 3 - 4 | 532 | 61 | ||
| XM_021467580.1 | XP_021323255.1 | 3 - 4 | 532 | 61 | ||
| GenBank mRNA | BC044337.1 | - | 3 - 4 | 61 | ||
| DQ323506.1 | - | 1 - 2 | 61 | |||
| EH440607.1 | - | 3 - 4 | 531 | 61 | ||
| U66568.1 | - | 3 - 4 | 61 | |||
More Information
Set Membership: |
|

mef2aa
Genomic Map