Assay Details
| Gene Symbol: mef2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131301.2 | NP_571376.2 | 5 - 6 | 861 | 87 | |
| XM_021467578.1 | XP_021323253.1 | 5 - 6 | 871 | 87 | ||
| XM_021467580.1 | XP_021323255.1 | 5 - 6 | 871 | 87 | ||
| GenBank mRNA | BC044337.1 | - | 5 - 6 | 861 | 87 | |
| U66568.1 | - | 5 - 6 | 811 | 87 | ||
More Information
Set Membership: |
|

mef2aa
Genomic Map