Assay Details
| Gene Symbol: mef2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131301.2 | NP_571376.2 | 6 - 7 | 1070 | 94 | |
| XM_005159044.4 | XP_005159101.1 | 7 - 8 | 1214 | 94 | ||
| XM_021467576.1 | XP_021323251.1 | 7 - 8 | 1215 | 94 | ||
| XM_021467579.1 | XP_021323254.1 | 6 - 7 | 1083 | 94 | ||
| XM_021467580.1 | XP_021323255.1 | 6 - 7 | 1080 | 94 | ||
| GenBank mRNA | BC044337.1 | - | 6 - 7 | 1076 | 94 | |
| EH466515.1 | - | 1 - 2 | 147 | 94 | ||
| U66568.1 | - | 6 - 7 | 1026 | 94 | ||
More Information
Set Membership: |
|

mef2aa
Genomic Map