Assay Details
| Gene Symbol: mef2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131301.2 | NP_571376.2 | 7 - 8 | 1257 | 61 | |
| XM_005159042.4 | XP_005159099.1 | 8 - 9 | 1404 | 61 | ||
| XM_005159044.4 | XP_005159101.1 | 8 - 9 | 1401 | 61 | ||
| XM_005159045.4 | XP_005159102.1 | 7 - 8 | 1272 | 61 | ||
| GenBank mRNA | BC044337.1 | - | 7 - 8 | 1263 | 61 | |
| DQ323506.1 | - | 5 - 6 | 802 | 61 | ||
| EB903808.1 | - | 2 - 3 | 270 | 61 | ||
| U66568.1 | - | 7 - 8 | 1213 | 61 | ||
More Information
Set Membership: |
|

mef2aa
Genomic Map