Assay Details
| Gene Symbol: mef2aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131301.2 | NP_571376.2 | 9 - 10 | 1511 | 58 | |
| XM_005159042.4 | XP_005159099.1 | 10 - 11 | 1658 | 58 | ||
| XM_005159044.4 | XP_005159101.1 | 10 - 11 | 1655 | 58 | ||
| XM_005159045.4 | XP_005159102.1 | 9 - 10 | 1526 | 58 | ||
| XM_021467574.1 | XP_021323249.1 | 11 - 12 | 1683 | 58 | ||
| XM_021467575.1 | XP_021323250.1 | 11 - 12 | 1684 | 58 | ||
| XM_021467576.1 | XP_021323251.1 | 11 - 12 | 1680 | 58 | ||
| XM_021467577.1 | XP_021323252.1 | 10 - 11 | 1551 | 58 | ||
| XM_021467578.1 | XP_021323253.1 | 10 - 11 | 1548 | 58 | ||
| XM_021467579.1 | XP_021323254.1 | 10 - 11 | 1548 | 58 | ||
| XM_021467580.1 | XP_021323255.1 | 10 - 11 | 1545 | 58 | ||
| GenBank mRNA | BC044337.1 | - | 9 - 10 | 1517 | 58 | |
| DQ323506.1 | - | 7 - 8 | 1056 | 58 | ||
| EB903808.1 | - | 4 - 5 | 524 | 58 | ||
| U66568.1 | - | 9 - 10 | 1467 | 58 | ||
More Information
Set Membership: |
|

mef2aa
Genomic Map