Assay Details
| Gene Symbol: notch1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131302.2 | NP_571377.2 | 12 - 13 | 2100 | 71 | |
| XM_005168649.4 | XP_005168706.1 | 11 - 12 | 2128 | 71 | ||
| GenBank mRNA | AY805407.1 | - | 12 - 13 | 2100 | 71 | |
More Information
Set Membership: |
|

notch1b
Genomic Map