Assay Details
| Gene Symbol: notch1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131302.2 | NP_571377.2 | 22 - 23 | 3721 | 85 | |
| XM_005168649.4 | XP_005168706.1 | 22 - 23 | 3869 | 85 | ||
| GenBank mRNA | AY805407.1 | - | 22 - 23 | 3721 | 85 | |
More Information
Set Membership: |
|

notch1b
Genomic Map