Assay Details
| Gene Symbol: notch1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131302.2 | NP_571377.2 | 26 - 27 | 5094 | 62 | |
| GenBank mRNA | AY805407.1 | - | 26 - 27 | 5094 | 62 | |
| Y10352.1 | - | 933 | 62 | |||
More Information
Set Membership: |
|

notch1b
Genomic Map