Assay Details
| Gene Symbol: ldb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131316.1 | NP_571391.1 | 10 - 11 | 1190 | 54 | |
| XM_009307285.2 | XP_009305560.1 | 12 - 13 | 1260 | 54 | ||
| XM_009307287.2 | XP_009305562.1 | 12 - 13 | 1245 | 54 | ||
| XM_017358692.2 | XP_017214181.1 | 12 - 13 | 1194 | 54 | ||
| GenBank mRNA | AF031378.1 | - | 10 - 11 | 1190 | 54 | |
More Information
Set Membership: |
|

ldb1a
Genomic Map