Assay Details
| Gene Symbol: ldb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131316.1 | NP_571391.1 | 3 - 4 | 366 | 126 | |
| XM_005156756.4 | XP_005156813.1 | 4 - 5 | 912 | 126 | ||
| XM_005156757.3 | XP_005156814.1 | 5 - 6 | 421 | 126 | ||
| XM_005156765.4 | XP_005156822.1 | 3 - 4 | 465 | 126 | ||
| XM_005156768.4 | XP_005156825.1 | 3 - 4 | 464 | 126 | ||
| XM_009307284.3 | XP_009305559.1 | 4 - 5 | 910 | 126 | ||
| XM_009307285.2 | XP_009305560.1 | 5 - 6 | 421 | 126 | ||
| XM_009307286.2 | XP_009305561.1 | 5 - 6 | 421 | 126 | ||
| XM_009307287.2 | XP_009305562.1 | 5 - 6 | 421 | 126 | ||
| XM_009307288.3 | XP_009305563.1 | 3 - 4 | 504 | 126 | ||
| XM_009307289.2 | XP_009305564.1 | 5 - 6 | 421 | 126 | ||
| XM_009307290.2 | XP_009305565.1 | 5 - 6 | 421 | 126 | ||
| XM_009307291.3 | XP_009305566.1 | 5 - 6 | 421 | 126 | ||
| XM_009307292.2 | XP_009305567.1 | 5 - 6 | 421 | 126 | ||
| XM_021480515.1 | XP_021336190.1 | 5 - 6 | 372 | 126 | ||
| XM_021480517.1 | XP_021336192.1 | 3 - 4 | 206 | 126 | ||
| GenBank mRNA | AB250382.1 | - | 2 - 3 | 75 | 126 | |
| AF031378.1 | - | 3 - 4 | 366 | 126 | ||
| BC097093.1 | - | 3 - 4 | 341 | 126 | ||
| BC164854.1 | - | 3 - 4 | 234 | 126 | ||
More Information
Set Membership: |
|

ldb1a
Genomic Map