Assay Details
| Gene Symbol: ldb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131316.1 | NP_571391.1 | 7 - 8 | 847 | 135 | |
| XM_005156768.4 | XP_005156825.1 | 7 - 8 | 945 | 135 | ||
| XM_009307286.2 | XP_009305561.1 | 9 - 10 | 902 | 135 | ||
| XM_009307287.2 | XP_009305562.1 | 9 - 10 | 902 | 135 | ||
| XM_009307291.3 | XP_009305566.1 | 9 - 10 | 902 | 135 | ||
| XM_009307292.2 | XP_009305567.1 | 9 - 10 | 902 | 135 | ||
| XM_017358691.2 | XP_017214180.1 | 9 - 10 | 851 | 135 | ||
| XM_017358692.2 | XP_017214181.1 | 9 - 10 | 851 | 135 | ||
| XM_017358693.2 | XP_017214182.1 | 9 - 10 | 849 | 135 | ||
| GenBank mRNA | AB250382.1 | - | 6 - 7 | 556 | 135 | |
| AF031378.1 | - | 7 - 8 | 847 | 135 | ||
| BC084686.1 | - | 7 - 8 | 655 | 135 | ||
| BC097093.1 | - | 7 - 8 | 822 | 135 | ||
| BC164854.1 | - | 7 - 8 | 715 | 135 | ||
More Information
Set Membership: |
|

ldb1a
Genomic Map