Assay Details
Gene Symbol: gap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131341.1 | NP_571416.1 | 2 - 3 | 709 | 82 | |
GenBank mRNA | BC076458.1 | - | 2 - 3 | 755 | 82 | |
BC165904.1 | - | 2 - 3 | 551 | 82 | ||
L27645.1 | - | 2 - 3 | 709 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
