Assay Details
| Gene Symbol: fabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131431.1 | NP_571506.1 | 1 - 2 | 78 | 71 | |
| XM_005170903.3 | XP_005170960.1 | 1 - 2 | 478 | 71 | ||
| GenBank mRNA | AF180921.2 | - | 1 - 2 | 74 | 71 | |
| AF541953.1 | - | 1 - 2 | 78 | 71 | ||
| AJ132590.1 | - | 1 - 2 | 78 | 71 | ||
| BC075970.1 | - | 1 - 2 | 82 | 71 | ||
More Information
Set Membership: |
|

fabp2
Genomic Map