Assay Details
| Gene Symbol: snap25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131435.1 | NP_571510.1 | 2 - 3 | 252 | 84 | |
| XM_005160623.4 | XP_005160680.1 | 2 - 3 | 279 | 84 | ||
| GenBank mRNA | BC059439.1 | - | 2 - 3 | 250 | 84 | |
| BC063942.1 | - | 2 - 3 | 252 | 84 | ||
More Information
Set Membership: |
|

snap25a
Genomic Map