Assay Details
| Gene Symbol: snap25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131435.1 | NP_571510.1 | 7 - 8 | 732 | 82 | |
| XM_005160623.4 | XP_005160680.1 | 7 - 8 | 759 | 82 | ||
| GenBank mRNA | AF091593.1 | - | 3 - 4 | 308 | 82 | |
| BC059439.1 | - | 7 - 8 | 730 | 82 | ||
| BC063942.1 | - | 7 - 8 | 732 | 82 | ||
More Information
Set Membership: |
|

snap25a
Genomic Map