Assay Details
| Gene Symbol: npy8br | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131436.1 | NP_571511.1 | 1 | 1046 | 69 | |
| XM_009304308.3 | XP_009302583.1 | 2 | 1377 | 69 | ||
| GenBank mRNA | BC162788.1 | - | 1 | 69 | ||
| BC162808.1 | - | 1 | 1109 | 69 | ||
More Information
Set Membership: |
|

npy8br
Genomic Map