Assay Details
| Gene Symbol: npy8ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131437.1 | NP_571512.1 | 1 | 749 | 61 | |
| GenBank mRNA | BC163700.1 | - | 1 | 831 | 61 | |
| BC163705.1 | - | 1 | 831 | 61 | ||
More Information
Set Membership: |
|

npy8ar
Genomic Map