Assay Details
| Gene Symbol: eif4e1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131454.1 | NP_571529.1 | 5 - 6 | 454 | 65 | |
| GenBank mRNA | AF176316.1 | - | 5 - 6 | 511 | 65 | |
| AF176317.1 | - | 5 - 6 | 454 | 65 | ||
| BC055649.1 | - | 5 - 6 | 423 | 65 | ||
More Information
Set Membership: |
|

eif4e1b
Genomic Map