Assay Details
| Gene Symbol: tbx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131506.2 | NP_571581.2 | 1 - 2 | 296 | 63 | |
| XM_021466746.1 | XP_021322421.1 | 1 - 2 | 486 | 63 | ||
| GenBank mRNA | AF239664.1 | - | 1 - 2 | 298 | 63 | |
| AF253325.1 | - | 1 - 2 | 311 | 63 | ||
| BC163475.1 | - | 1 - 2 | 287 | 63 | ||
More Information
Set Membership: |
|

tbx20
Genomic Map