Assay Details
| Gene Symbol: mxtx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131560.3 | NP_571635.2 | 2 - 3 | 279 | 83 | |
| GenBank mRNA | AB034245.1 | - | 2 - 3 | 265 | 83 | |
| BC127384.1 | - | 2 - 3 | 280 | 83 | ||
More Information
Set Membership: |
|

mxtx1
Genomic Map