Assay Details
| Gene Symbol: prph2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131567.1 | NP_571642.1 | 2 - 3 | 1174 | 83 | |
| GenBank mRNA | BC059645.1 | - | 2 - 3 | 1174 | 83 | |
| BC155196.1 | - | 2 - 3 | 1173 | 83 | ||
More Information
Set Membership: |
|

prph2b
Genomic Map