Assay Details
| Gene Symbol: adarb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131610.3 | NP_571685.2 | 5 - 6 | 1286 | 74 | |
| XM_021469375.1 | XP_021325050.1 | 6 - 7 | 1347 | 74 | ||
| GenBank mRNA | AF403110.1 | - | 4 - 5 | 1217 | 74 | |
| AF403111.1 | - | 4 - 5 | 1264 | 74 | ||
| AF403112.1 | - | 5 - 6 | 1275 | 74 | ||
| AF403113.1 | - | 5 - 6 | 1322 | 74 | ||
More Information
Set Membership: |
|

adarb1a
Genomic Map