Assay Details
| Gene Symbol: adarb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131610.3 | NP_571685.2 | 6 - 7 | 1399 | 75 | |
| XM_021469375.1 | XP_021325050.1 | 7 - 8 | 1460 | 75 | ||
| GenBank mRNA | AF124333.1 | - | 1 - 2 | 78 | 75 | |
| AF403110.1 | - | 5 - 6 | 1330 | 75 | ||
| AF403111.1 | - | 5 - 6 | 1377 | 75 | ||
| AF403112.1 | - | 6 - 7 | 1388 | 75 | ||
| AF403113.1 | - | 6 - 7 | 1435 | 75 | ||
More Information
Set Membership: |
|

adarb1a
Genomic Map