Assay Details
| Gene Symbol: eif4ea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131733.1 | NP_571808.1 | 7 - 8 | 664 | 131 | |
| GenBank mRNA | AF257519.1 | - | 7 - 8 | 664 | 131 | |
| BC071364.1 | - | 7 - 8 | 677 | 131 | ||
More Information
Set Membership: |
|

eif4ea
Genomic Map