Assay Details
| Gene Symbol: nkx2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131776.1 | NP_571851.1 | 1 - 2 | 492 | 66 | |
| GenBank mRNA | AF321112.1 | - | 1 - 2 | 492 | 66 | |
| BC162657.1 | - | 1 - 2 | 487 | 66 | ||
| BC162660.1 | - | 1 - 2 | 487 | 66 | ||
More Information
Set Membership: |
|

nkx2.1
Genomic Map