Assay Details
| Gene Symbol: anxa11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181765.2 | NP_861430.2 | 3 - 4 | 377 | 116 | |
| GenBank mRNA | AY178801.1 | - | 3 - 4 | 369 | 116 | |
More Information
Set Membership: |
|

anxa11a
Genomic Map