Assay Details
| Gene Symbol: anxa11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181765.2 | NP_861430.2 | 8 - 9 | 1089 | 98 | |
| NM_183410.2 | NP_899670.1 | 8 - 9 | 960 | 98 | ||
| GenBank mRNA | AY178801.1 | - | 8 - 9 | 1081 | 98 | |
| BC053208.1 | - | 8 - 9 | 922 | 98 | ||
More Information
Set Membership: |
|

anxa11a
Genomic Map