Assay Details
| Gene Symbol: f9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182868.2 | NP_878288.2 | 2 - 3 | 317 | 71 | |
| GenBank mRNA | AF515270.1 | - | 2 - 3 | 245 | 71 | |
| EH506608.1 | - | 2 - 3 | 260 | 71 | ||
More Information
Set Membership: |
|

f9a
Genomic Map