Assay Details
| Gene Symbol: f9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182868.2 | NP_878288.2 | 7 - 8 | 1022 | 108 | |
| XM_005173201.4 | XP_005173258.1 | 5 - 6 | 755 | 108 | ||
| GenBank mRNA | AF515270.1 | - | 7 - 8 | 950 | 108 | |
| BC094987.1 | - | 4 - 5 | 482 | 108 | ||
| BC164757.1 | - | 4 - 5 | 486 | 108 | ||
| EH490346.1 | - | 5 - 6 | 751 | 108 | ||
| EH516696.1 | - | 1 - 2 | 695 | 108 | ||
More Information
Set Membership: |
|

f9a
Genomic Map