Assay Details
| Gene Symbol: tgfb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182873.1 | NP_878293.1 | 6 - 7 | 1252 | 80 | |
| GenBank mRNA | AY178450.1 | - | 6 - 7 | 1252 | 80 | |
| BC162361.1 | - | 6 - 7 | 1161 | 80 | ||
| BC162366.1 | - | 6 - 7 | 1161 | 80 | ||
More Information
Set Membership: |
|

tgfb1a
Genomic Map