Assay Details
| Gene Symbol: slc4a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198338.1 | NP_938152.1 | 14 - 15 | 1686 | 142 | |
| GenBank mRNA | AF350072.1 | - | 14 - 15 | 1686 | 142 | |
| AY098936.1 | - | 14 - 15 | 1719 | 142 | ||
| BC093126.1 | - | 7 - 8 | 987 | 142 | ||
| BC163932.1 | - | 14 - 15 | 1672 | 142 | ||
More Information
Set Membership: |
|

slc4a1a
Genomic Map