Assay Details
| Gene Symbol: slc4a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198338.1 | NP_938152.1 | 20 - 21 | 2783 | 78 | |
| GenBank mRNA | AF350072.1 | - | 20 - 21 | 2783 | 78 | |
| AY098936.1 | - | 20 - 21 | 2816 | 78 | ||
| BC124121.1 | - | 6 - 7 | 791 | 78 | ||
| BC134843.1 | - | 7 - 8 | 1116 | 78 | ||
| BC152535.1 | - | 5 - 6 | 815 | 78 | ||
| BC163932.1 | - | 20 - 21 | 2769 | 78 | ||
More Information
Set Membership: |
|

slc4a1a
Genomic Map