Assay Details
| Gene Symbol: clic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199524.1 | NP_955818.1 | 1 - 2 | 259 | 67 | |
| XM_005161114.4 | XP_005161171.1 | 2 - 3 | 584 | 67 | ||
| GenBank mRNA | BC062860.1 | - | 1 - 2 | 276 | 67 | |
More Information
Set Membership: |
|

clic3
Genomic Map