Assay Details
| Gene Symbol: phgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199577.2 | NP_955871.2 | 10 - 11 | 1276 | 64 | |
| GenBank mRNA | BC056334.1 | - | 10 - 11 | 1232 | 64 | |
| EH460654.1 | - | 2 - 3 | 608 | 64 | ||
| EH609675.1 | - | 2 - 3 | 608 | 64 | ||
More Information
Set Membership: |
|

phgdh
Genomic Map