Assay Details
| Gene Symbol: dbi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199608.2 | NP_955902.1 | 3 - 4 | 303 | 83 | |
| GenBank mRNA | BC045916.1 | - | 3 - 4 | 238 | 83 | |
| BC062845.1 | - | 3 - 4 | 223 | 83 | ||
| BC164432.1 | - | 219 | 83 | |||
| CK028074.1 | - | 3 - 4 | 223 | 83 | ||
| EH546049.1 | - | 3 - 4 | 338 | 83 | ||
More Information
Set Membership: |
|

dbi
Genomic Map