Assay Details
| Gene Symbol: tomm40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199614.1 | NP_955908.1 | 6 - 7 | 614 | 66 | |
| XM_005158091.4 | XP_005158148.1 | 6 - 7 | 1022 | 66 | ||
| XM_017351318.2 | XP_017206807.1 | 6 - 7 | 811 | 66 | ||
| GenBank mRNA | BC053295.1 | - | 6 - 7 | 614 | 66 | |
More Information
Set Membership: |
|

tomm40
Genomic Map