Assay Details
| Gene Symbol: slc43a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327986.1 | NP_001314915.1 | 2 - 3 | 460 | 103 | |
| GenBank mRNA | BC044509.1 | - | 2 - 3 | 425 | 103 | |
| BC164469.1 | - | 1 - 2 | 182 | 103 | ||
More Information
Set Membership: |
|

slc43a1a
Genomic Map