Assay Details
| Gene Symbol: slc43a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327986.1 | NP_001314915.1 | 5 - 6 | 780 | 69 | |
| GenBank mRNA | BC044509.1 | - | 5 - 6 | 745 | 69 | |
| BC164469.1 | - | 4 - 5 | 502 | 69 | ||
More Information
Set Membership: |
|

slc43a1a
Genomic Map