Assay Details
| Gene Symbol: slc43a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001327986.1 | NP_001314915.1 | 7 - 8 | 990 | 98 | |
| GenBank mRNA | BC044509.1 | - | 7 - 8 | 955 | 98 | |
| BC164469.1 | - | 6 - 7 | 712 | 98 | ||
More Information
Set Membership: |
|

slc43a1a
Genomic Map