Assay Details
| Gene Symbol: rbm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199925.1 | NP_956219.1 | 4 - 5 | 496 | 68 | |
| GenBank mRNA | BC049507.1 | - | 4 - 5 | 496 | 68 | |
| BC065641.1 | - | 4 - 5 | 518 | 68 | ||
More Information
Set Membership: |
|

rbm7
Genomic Map