Assay Details
| Gene Symbol: plpp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199953.1 | NP_956247.1 | 5 - 6 | 1098 | 124 | |
| GenBank mRNA | BC054702.1 | - | 5 - 6 | 1098 | 124 | |
More Information
Set Membership: |
|

plpp2b
Genomic Map