Assay Details
| Gene Symbol: fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199955.1 | NP_956249.1 | 2 - 3 | 173 | 65 | |
| GenBank mRNA | BC053266.1 | - | 2 - 3 | 173 | 65 | |
More Information
Set Membership: |
|

fgfr1op2
Genomic Map