Assay Details
| Gene Symbol: rpl7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200047.1 | NP_956341.1 | 3 - 4 | 418 | 102 | |
| GenBank mRNA | BC059533.1 | - | 3 - 4 | 418 | 102 | |
| BC071352.1 | - | 4 - 5 | 434 | 102 | ||
More Information
Set Membership: |
|

rpl7a
Genomic Map