Assay Details
| Gene Symbol: dcaf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200069.1 | NP_956363.1 | 1 - 2 | 280 | 60 | |
| XM_005163996.3 | XP_005164053.1 | 1 - 2 | 521 | 60 | ||
| GenBank mRNA | BC053157.1 | - | 1 - 2 | 280 | 60 | |
More Information
Set Membership: |
|

dcaf7
Genomic Map