Assay Details
| Gene Symbol: pip4k2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200101.2 | NP_956395.2 | 1 - 2 | 163 | 71 | |
| GenBank mRNA | BC045883.1 | - | 1 - 2 | 300 | 71 | |
| BC071466.1 | - | 1 - 2 | 163 | 71 | ||
More Information
Set Membership: |
|

pip4k2ca
Genomic Map