Assay Details
| Gene Symbol: pip4k2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200101.2 | NP_956395.2 | 8 - 9 | 1078 | 91 | |
| GenBank mRNA | BC045883.1 | - | 8 - 9 | 1215 | 91 | |
| BC071466.1 | - | 8 - 9 | 1078 | 91 | ||
More Information
Set Membership: |
|

pip4k2ca
Genomic Map